Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Bis(trifluoromethyl)benzyl alcohol, 98%

$57.27 - $229.35

Chemical Identifiers

Chemical Name or Material 2,4-Bis(trifluoromethyl)benzyl alcohol
CAS 143158-15-0
Molecular Formula C9H6F6O
Formula Weight 244.15
MDL Number MFCD00042491
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2075703

 
alfa aesar™
B2075703
1g Each for $57.27

AAB2075706

 
alfa aesar™
B2075706
5g Each for $229.35
Description & Specifications

Specifications

Formula Weight 244.15
Chemical Name or Material 2,4-Bis(trifluoromethyl)benzyl alcohol
Assay 98%
Molecular Formula C9H6F6O
MDL Number MFCD00042491
CAS 143158-15-0
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.