Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Bis(trifluoromethyl)benzoic acid, 98%

$50.34 - $207.93

Chemical Identifiers

Chemical Name or Material 2,4-Bis(trifluoromethyl)benzoic acid
CAS 32890-87-2
Molecular Formula C9H4F6O2
Formula Weight 258.12
MDL Number MFCD00042496
Beilstein 2057460
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2073103

  SDS
alfa aesar™
B2073103
1g Each for $50.34

AAB2073106

  SDS
alfa aesar™
B2073106
5g Each for $207.93
Description & Specifications

Specifications

Formula Weight 258.12
Chemical Name or Material 2,4-Bis(trifluoromethyl)benzoic acid
Assay 98%
Molecular Formula C9H4F6O2
MDL Number MFCD00042496
CAS 32890-87-2
Beilstein 2057460
Melting Point 108° to 110°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.