missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Bis(trifluoromethyl)benzaldehyde, TCI America™

$50.42 - $161.95

Chemical Identifiers

CAS 59664-42-5
Molecular Formula C9H4F6O
Molecular Weight (g/mol) 242.12
MDL Number MFCD00083535
InChI Key DLRCWXOCZIUZBS-UHFFFAOYSA-N
Synonym 2,4-bis trifluoromethyl benzaldehyde, 2,4-ditrifluoromethylbenzaldehyde, 2,4-bis-trifluoromethyl-benzaldehyde, benzaldehyde, 2,4-bis trifluoromethyl, pubchem7129, acmc-209xb0, ksc610e6l, 2,4-di trifluoromethyl benzaldehyde, 2,4-bis trifluoromethyl benzaldehyde, 2,4-bis trifluoromethyl-benzaldehyde
PubChem CID 2736095
IUPAC Name 2,4-bis(trifluoromethyl)benzaldehyde
SMILES C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B42061G View Documents TCI America
B42061G
1g Each for $50.42
 
B42065G View Documents TCI America
B42065G
5g Each for $161.95
 
Specifications

Chemical Identifiers

59664-42-5
242.12
DLRCWXOCZIUZBS-UHFFFAOYSA-N
2736095
C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)C=O
C9H4F6O
MFCD00083535
2,4-bis trifluoromethyl benzaldehyde, 2,4-ditrifluoromethylbenzaldehyde, 2,4-bis-trifluoromethyl-benzaldehyde, benzaldehyde, 2,4-bis trifluoromethyl, pubchem7129, acmc-209xb0, ksc610e6l, 2,4-di trifluoromethyl benzaldehyde, 2,4-bis trifluoromethyl benzaldehyde, 2,4-bis trifluoromethyl-benzaldehyde
2,4-bis(trifluoromethyl)benzaldehyde

Specifications

172°C
1g
C9H4F6O
2,4-bis trifluoromethyl benzaldehyde, 2,4-ditrifluoromethylbenzaldehyde, 2,4-bis-trifluoromethyl-benzaldehyde, benzaldehyde, 2,4-bis trifluoromethyl, pubchem7129, acmc-209xb0, ksc610e6l, 2,4-di trifluoromethyl benzaldehyde, 2,4-bis trifluoromethyl benzaldehyde, 2,4-bis trifluoromethyl-benzaldehyde
C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)C=O
242.12
242.12
≥97.0% (GC)
Yellow
59664-42-5
MFCD00083535
DLRCWXOCZIUZBS-UHFFFAOYSA-N
2,4-bis(trifluoromethyl)benzaldehyde
2736095
Clear Liquid at 20°C
2,4-Bis(trifluoromethyl)benzaldehyde
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator