Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Bis(trifluoromethyl)benzaldehyde, 97%

$63.14 - $185.34

Chemical Identifiers

Chemical Name or Material 2,4-Bis(trifluoromethyl)benzaldehyde
CAS 59664-42-5
Molecular Formula C9H4F6O
Formula Weight 242.12
MDL Number MFCD00083535
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2097803

  SDS
alfa aesar™
B2097803
1g Each for $63.14

AAB2097806

  SDS
alfa aesar™
B2097806
5g Each for $185.34
Description & Specifications

Specifications

Formula Weight 242.12
Assay 97%
Boiling Point 172°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Quantity 1g
CAS 59664-42-5
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C9H4F6O
Flash Point 68°C (155°F)
MDL Number MFCD00083535
Chemical Name or Material 2,4-Bis(trifluoromethyl)benzaldehyde
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.