Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',4'-Bis(trifluoromethyl)acetophenone, 97%

$68.70 - $164.24

Chemical Identifiers

Chemical Name or Material 2',4'-Bis(trifluoromethyl)acetophenone
CAS 237069-82-8
Molecular Formula C10H6F6O
Formula Weight 256.15
MDL Number MFCD00042495
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2680203

  SDS
alfa aesar™
H2680203
1g Each for $164.24

AAH26802MD

  SDS
alfa aesar™
H26802MD
250mg Each for $68.70
Description & Specifications

Specifications

Formula Weight 256.15
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Molecular Formula C10H6F6O
MDL Number MFCD00042495
CAS 237069-82-8
Chemical Name or Material 2',4'-Bis(trifluoromethyl)acetophenone
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.