missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Biphenylyl)-2-propanol, 95%

$67.85 - $280.66

Chemical Identifiers

CAS 34352-74-4
Molecular Formula C15H16O
Molecular Weight (g/mol) 212.292
MDL Number MFCD00014390
InChI Key GOKGIYHIVSGXDM-UHFFFAOYSA-N
Synonym 2-4-biphenylyl-2-propanol, 2-4-phenylphenyl propan-2-ol, 2-biphenyl-4-yl propan-2-ol, 2-1,1'-biphenyl-4-yl propan-2-ol, 2-4-biphenyl-2-propanol, 1,1'-biphenyl-4-methanol, .alpha.,.alpha.-dimethyl, 2-1,1'-biphenyl-4-yl propan-2-ol, acmc-1aevv, cambridge id 5135301, 1,1'-biphenyl-4-methanol, alpha,alpha-dimethyl
PubChem CID 118654
IUPAC Name 2-(4-phenylphenyl)propan-2-ol
SMILES CC(C)(C1=CC=C(C=C1)C2=CC=CC=C2)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0136409 View Documents Alfa Aesar™
L0136409
10g Each for $67.85
 
AAL0136418 View Documents Alfa Aesar™
L0136418
50g Each for $280.66
 
Specifications

Chemical Identifiers

34352-74-4
212.292
GOKGIYHIVSGXDM-UHFFFAOYSA-N
118654
CC(C)(C1=CC=C(C=C1)C2=CC=CC=C2)O
C15H16O
MFCD00014390
2-4-biphenylyl-2-propanol, 2-4-phenylphenyl propan-2-ol, 2-biphenyl-4-yl propan-2-ol, 2-1,1'-biphenyl-4-yl propan-2-ol, 2-4-biphenyl-2-propanol, 1,1'-biphenyl-4-methanol, .alpha.,.alpha.-dimethyl, 2-1,1'-biphenyl-4-yl propan-2-ol, acmc-1aevv, cambridge id 5135301, 1,1'-biphenyl-4-methanol, alpha,alpha-dimethyl
2-(4-phenylphenyl)propan-2-ol

Specifications

90°C to 93°C
34352-74-4
MFCD00014390
2-4-biphenylyl-2-propanol, 2-4-phenylphenyl propan-2-ol, 2-biphenyl-4-yl propan-2-ol, 2-1,1'-biphenyl-4-yl propan-2-ol, 2-4-biphenyl-2-propanol, 1,1'-biphenyl-4-methanol, .alpha.,.alpha.-dimethyl, 2-1,1'-biphenyl-4-yl propan-2-ol, acmc-1aevv, cambridge id 5135301, 1,1'-biphenyl-4-methanol, alpha,alpha-dimethyl
CC(C)(C1=CC=C(C=C1)C2=CC=CC=C2)O
212.292
212.29
2-(4-Biphenylyl)-2-propanol
10g
C15H16O
1947575
GOKGIYHIVSGXDM-UHFFFAOYSA-N
2-(4-phenylphenyl)propan-2-ol
118654
97%
Safety and Handling

Safety and Handling

TSCA : TSCA