missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-aminophenylthio)acetic Acid 98%, ACROS Organics™

$49.56 - $49.56

Chemical Identifiers

CAS 104-18-7
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C8 H9 N O2 S
Linear Formula NH2C6H4SCH2COOH
Formula Weight 183.23
MDL Number MFCD00025360
Chemical Name or Material 2-(4-Aminophenylthio)acetic acid
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC382030010 SDS Thermo Fisher Scientific
AC382030010
Glass bottle 1g Each for $49.56
AC382030050 SDS Thermo Fisher Scientific
AC382030050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

104-18-7
100.0
C8 H9 N O2 S
183.23
2-(4-Aminophenylthio)acetic acid
97.5
98%
NH2C6H4SCH2COOH
MFCD00025360
≥97.5%

Specifications

104-18-7
97.5
Glass bottle
193° to 203°C
1g
C8 H9 N O2 S
183.23
2-(4-Aminophenylthio)acetic acid
Authentic
100.0
Light khaki
Crystalline Powder
98%
NH2C6H4SCH2COOH
MFCD00025360
≥97.5%

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning