Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Aminophenyl)-4-(4-methoxyphenyl)thiazole, 97%

Chemical Identifiers

Chemical Name or Material 2-(4-Aminophenyl)-4-(4-methoxyphenyl)thiazole
CAS 749151-25-5
Formula Weight 282.36
Synonym 4-[4-(4-Methoxyphenyl)-2-thiazolyl]aniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6645803

 
alfa aesar™
H6645803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66458MD

 
alfa aesar™
H66458MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 282.36
Chemical Name or Material 2-(4-Aminophenyl)-4-(4-methoxyphenyl)thiazole
Assay 97%
Physical Form Crystalline Solid
Synonym 4-[4-(4-Methoxyphenyl)-2-thiazolyl]aniline
Color Brown
Quantity 1g
CAS 749151-25-5