Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol, 96%

$477.65 - $477.65

Chemical Identifiers

Chemical Name or Material 2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol
CAS 722-92-9
Molecular Formula C9H7F6NO
Formula Weight 259.15
MDL Number MFCD00039258
Beilstein 1473737
Synonym 4-Amino-alpha-alpha-bis(trifluoromethyl)benzyl alcohol; 4-(Hexafluoro-2-hydroxyisopropyl)aniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2404814

  SDS
alfa aesar™
B2404814
25g Each for $477.65
Description & Specifications

Specifications

Formula Weight 259.15
Beilstein 1473737
Synonym 4-Amino-alpha-alpha-bis(trifluoromethyl)benzyl alcohol; 4-(Hexafluoro-2-hydroxyisopropyl)aniline
Health Hazard 1 Danger
Quantity 25g
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 722-92-9
Melting Point 147° to 152°C
MDL Number MFCD00039258
Assay 96%
Chemical Name or Material 2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol
Molecular Formula C9H7F6NO
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.