Please login to your online account to display your discounted pricing

2,4,8,10-Tetraoxaspiro[5.5]undecane, 98+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2,4,8,10-Tetraoxaspiro[5.5]undecane
Name Note 98+%
CAS 126-54-5
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C7H12O4
Formula Weight 160.17
MDL Number MFCD00006565
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC205350050

  SDS
thermo fisher scientific
AC205350050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Formula Weight 160.17
Physical Form Crystalline Mass
Quantity 5g
Color White
Melting Point 48° to 52°C
TSCA TSCA
EINECS Number 204-792-0
CAS Min % 98.0
CAS Max % 100.0
Beilstein 19, V,11, 342
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym Pentaerythritol diformal
Health Hazard 1 HYGROSCOPIC
CAS 126-54-5
Molecular Formula C7H12O4
Assay Percent Range 98+%
Chemical Name or Material 2,4,8,10-Tetraoxaspiro[5.5]undecane
MDL Number MFCD00006565
Name Note 98+%
Safety and Handling

HYGROSCOPIC