Please login to your online account to display your discounted pricing

2,4,8,10-Tetraoxaspiro[5.5]undecane, 98+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC205350050

 View more versions of this product

Catalog No. AC205350050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Formula Weight 160.17
Physical Form Crystalline Mass
Quantity 5g
Color White
Melting Point 48° to 52°C
TSCA TSCA
EINECS Number 204-792-0
CAS Min % 98.0
CAS Max % 100.0
Beilstein 19, V,11, 342
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym Pentaerythritol diformal
Health Hazard 1 HYGROSCOPIC
CAS 126-54-5
Molecular Formula C7H12O4
Assay Percent Range 98+%
Chemical Name or Material 2,4,8,10-Tetraoxaspiro[5.5]undecane
MDL Number MFCD00006565
Name Note 98+%

Specifications

Chemical Name or Material 2,4,8,10-Tetraoxaspiro[5.5]undecane
Name Note 98+%
CAS 126-54-5
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C7H12O4
Formula Weight 160.17
MDL Number MFCD00006565
Synonym Pentaerythritol diformal
Accessories

HYGROSCOPIC

SDS & Safety Documents