Please login to your online account to display your discounted pricing

2,4,6-tris(tert-butylperoxy)-1,3,5-triazine, Acros Organics


Quantity 1g

Manufacturer: thermo fisher scientific  AC378580010

 View more versions of this product

Catalog No. AC378580010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 345.4
Quantity 1g
Color Clear colorless
Assay 33 to 37 wt%
CAS 108-88-3
CAS 103-23-1
Chemical Name or Material 2,4,6-Tris(tert-butylperoxy)-1,3,5-triazine
Molecular Formula C15H27N3O6
CAS 854-44-4
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2,4,6-Tris(tert-butylperoxy)-1,3,5-triazine
CAS 854-44-4
CAS 108-88-3
CAS 103-23-1
Assay 33 to 37 wt%
Molecular Formula C15H27N3O6
Formula Weight 345.4
Accessories

Danger

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents