Please login to your online account to display your discounted pricing

2,4,6-tris(tert-butylperoxy)-1,3,5-triazine, Acros Organics

Chemical Identifiers

Chemical Name or Material 2,4,6-Tris(tert-butylperoxy)-1,3,5-triazine
CAS 854-44-4
CAS 108-88-3
CAS 103-23-1
Assay 33 to 37 wt%
Molecular Formula C15H27N3O6
Formula Weight 345.4
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC378580010

  SDS
thermo fisher scientific
AC378580010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 345.4
Quantity 1g
Color Clear colorless
Assay 33 to 37 wt%
CAS 103-23-1
CAS 108-88-3
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C15H27N3O6
Chemical Name or Material 2,4,6-Tris(tert-butylperoxy)-1,3,5-triazine
CAS 854-44-4
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.

Radical initiators