Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine, Mass Spec Std

$77.46 - $247.11

Chemical Identifiers

Chemical Name or Material 2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine
CAS 21674-38-4
Name Note Mass Spec Standard
Molecular Formula C24F45N3
Formula Weight 1185.22
MDL Number MFCD00042439
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL16678ME

  SDS
alfa aesar™
L16678ME
500mg Each for $247.11

AAL16678MC

  SDS
alfa aesar™
L16678MC
100mg Each for $77.46
Description & Specifications

Specifications

Formula Weight 1185.22
Chemical Name or Material 2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine
Melting Point 29° to 31°C
Boiling Point 150°C (1.5mmHg)
Quantity 500mg
Name Note Mass Spec Standard
CAS 21674-38-4
Molecular Formula C24F45N3
MDL Number MFCD00042439

Applications
2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine is used as a mass spectrometry standard.

Solubility
Insoluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.