Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine, Mass Spec Std


Quantity 100mg

Manufacturer: alfa aesar™  L16678MC

 View more versions of this product

Catalog No. AAL16678MC

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 1185.22
Chemical Name or Material 2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine
Molecular Formula C24F45N3
CAS 21674-38-4
Quantity 100mg
Name Note Mass Spec Standard
MDL Number MFCD00042439
Melting Point 29° to 31°C
Boiling Point 150°C (1.5mmHg)

Specifications

Chemical Name or Material 2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine
CAS 21674-38-4
Name Note Mass Spec Standard
Molecular Formula C24F45N3
Formula Weight 1185.22
MDL Number MFCD00042439

Applications
2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine is used as a mass spectrometry standard.

Solubility
Insoluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.