missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Tris[bis(methoxymethyl)amino]-1,3,5-triazine, TCI America™

Manufacturer:  TCI America T205925G

 View more versions of this product

Catalog No. T205925G



Specifications

Specifications

2,4,6-Tris[bis(methoxymethyl)amino]-1,3,5-triazine
52°C
3089-11-0
MFCD00047342
BNCADMBVWNPPIZ-UHFFFAOYSA-N
2-N,2-N,4-N,4-N,6-N,6-N-hexakis(methoxymethyl)-1,3,5-triazine-2,4,6-triamine
62479
Crystal-Powder at 20°C
White
25g
C15H30N6O6
N,N,N′,N′,N′′,N′′-Hexakis(methoxymethyl)melamine
COCN(COC)C1=NC(=NC(=N1)N(COC)COC)N(COC)COC
390.441
390.44
≥98.0% (GC)

Chemical Identifiers

3089-11-0
390.441
BNCADMBVWNPPIZ-UHFFFAOYSA-N
62479
COCN(COC)C1=NC(=NC(=N1)N(COC)COC)N(COC)COC
C15H30N6O6
MFCD00047342
N,N,N′,N′,N′′,N′′-Hexakis(methoxymethyl)melamine
2-N,2-N,4-N,4-N,6-N,6-N-hexakis(methoxymethyl)-1,3,5-triazine-2,4,6-triamine
Safety and Handling

Safety and Handling

EINECSNumber : (5)-1012&(5)-3839

RTECSNumber : XZ1065000

TSCA : Yes

Documents