Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Tris(allyloxy)-1,3,5-triazine, 98%, stab. with 100ppm hydroquinone

Chemical Identifiers

Chemical Name or Material 2,4,6-Tris(allyloxy)-1,3,5-triazine
CAS 101-37-1
Name Note stabilized with 100ppm hydroquinone
Molecular Formula C12H15N3O3
Formula Weight 249.27
MDL Number MFCD00006049
UN Number UN3077
Synonym Cyanuric acid triallyl ester; Triallyl cyanurate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1627418

 
alfa aesar™
L1627418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 249.27
Chemical Name or Material 2,4,6-Tris(allyloxy)-1,3,5-triazine
CAS 101-37-1
Health Hazard 2 GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Name Note stabilized with 100ppm hydroquinone
Assay 98%
Molecular Formula C12H15N3O3
MDL Number MFCD00006049
UN Number UN3077
Synonym Cyanuric acid triallyl ester; Triallyl cyanurate
Melting Point 25° to 28°C
Boiling Point 149° to 150°C (4mmHg)
Flash Point 166°C (331°F)
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number XZ2080000
TSCA TSCA
Health Hazard 1 Danger
Quantity 50g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.