missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Tris(3-pyridyl)-1,3,5-triazine, 97%

Manufacturer:  Alfa Aesar™ H3605203

 View more versions of this product

Catalog No. AAH3605203


 

Specifications

Specifications

2,4,6-Tris(3-pyridyl)-1,3,5-triazine
97%
MFCD23379843
UOSYDSDIRAXTTF-UHFFFAOYSA-N
2,4,6-tripyridin-3-yl-1,3,5-triazine
11472427
Solid
>300°C
42333-76-6
C18H12N6
2,4,6-Tri-3-pyridinyl-1,3,5-triazine
C1=CC(=CN=C1)C2=NC(=NC(=N2)C3=CN=CC=C3)C4=CN=CC=C4
312.336
312.33
White through Yellow
1g

Chemical Identifiers

42333-76-6
312.336
UOSYDSDIRAXTTF-UHFFFAOYSA-N
11472427
C18H12N6
MFCD23379843
2,4,6-Tri-3-pyridinyl-1,3,5-triazine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents