missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4,6-Tris(2,4,6-tribromophenoxy)-1,3,5-triazine 98.0+%, TCI America™
Supplier: TCI America T287125G
Specifications
2,4,6-Tris(2,4,6-tribromophenoxy)-1,3,5-triazine | |
230°C | |
C21H6Br9N3O3 | |
25 g | |
BDFBPPCACYFGFA-UHFFFAOYSA-N | |
2,4,6-tris(2,4,6-tribromophenoxy)-1,3,5-triazine | |
91820 | |
≥98.0% (HPLC,N) |
25713-60-4 | |
White | |
MFCD03092948 | |
Cyanuric Acid Tris(2,4,6-tribromophenyl) Ester, Tris(2,4,6-tribromophenyl) Cyanurate | |
C1=C(C=C(C(=C1Br)OC2=NC(=NC(=N2)OC3=C(C=C(C=C3Br)Br)Br)OC4=C(C=C(C=C4Br)Br)Br)Br)Br | |
1067.433 | |
1067.43 | |
Crystalline Powder |
Chemical Identifiers
25713-60-4 | |
1067.433 | |
BDFBPPCACYFGFA-UHFFFAOYSA-N | |
91820 | |
C1=C(C=C(C(=C1Br)OC2=NC(=NC(=N2)OC3=C(C=C(C=C3Br)Br)Br)OC4=C(C=C(C=C4Br)Br)Br)Br)Br |
C21H6Br9N3O3 | |
MFCD03092948 | |
Cyanuric Acid Tris(2,4,6-tribromophenyl) Ester, Tris(2,4,6-tribromophenyl) Cyanurate | |
2,4,6-tris(2,4,6-tribromophenoxy)-1,3,5-triazine |
Safety and Handling
EINECSNumber : (5)-6312
TSCA : Yes