missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Triphenylpyrylium tetrafluoroborate ≥97%, Acros Organics

Chemical Identifiers

CAS 448-61-3
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97+%
Molecular Formula C23H17BF4O
Formula Weight 396.19
MDL Number MFCD00012001
Chemical Name or Material 2, 4, 6-Triphenylpyrylium tetrafluoroborate
Assay ≥97.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC389290250 SDS Thermo Fisher Scientific
AC389290250
Plastic bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

448-61-3
100.0
C23H17BF4O
MFCD00012001
≥97.0%
97.0
97+%
396.19
2, 4, 6-Triphenylpyrylium tetrafluoroborate

Specifications

Crystalline Powder
25g
100.0
448-61-3
MFCD00012001
Plastic bottle
2, 4, 6-Triphenylpyrylium tetrafluoroborate
396.19
Yellow
97.0
C23H17BF4O
Authentic
97+%
≥97.0%

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning