missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trimethylpyridinium p-toluenesulfonate, 98%

Manufacturer:  Alfa Aesar™ H5521814

 View more versions of this product

Catalog No. AAH5521814



Specifications

Specifications

2,4,6-Trimethylpyridinium p-toluenesulfonate
25g
C15H19NO3S
293.38
128°C to 130°C
59229-09-3
MFCD00012810
98%

Chemical Identifiers

59229-09-3
C15H19NO3S
MFCD00012810
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents