missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Trimethylphenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1095803

 View more versions of this product

Catalog No. AAL1095803



Description

Description

2,4,6-Trimethylphenylhydrazine hydrochloride is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylphenylhydrazine hydrochloride is used as pharmaceutical intermediates.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications

Specifications

2,4,6-Trimethylphenylhydrazine hydrochloride
∽185°C (decomposition)
MFCD00052268
UN2811
mesitylhydrazine hydrochloride, 2,4,6-trimethylphenylhydrazine hydrochloride, 1-mesitylhydrazine hydrochloride, 2,4,6-trimethylphenyl hydrazine hydrochloride, hydrazine, 2,4,6-trimethylphenyl-, hydrochloride, mesityl hydrazine hydrochloride
XFGPCULNJGZRTH-UHFFFAOYSA-N
(2,4,6-trimethylphenyl)hydrazine;hydrochloride
2774883
97%
76195-82-9
C9H15ClN2
1 g
3700289
Soluble in water.
Cl.CC1=CC(C)=C(NN)C(C)=C1
186.68
186.69

Chemical Identifiers

76195-82-9
186.68
XFGPCULNJGZRTH-UHFFFAOYSA-N
2774883
Cl.CC1=CC(C)=C(NN)C(C)=C1
C9H15ClN2
MFCD00052268
mesitylhydrazine hydrochloride, 2,4,6-trimethylphenylhydrazine hydrochloride, 1-mesitylhydrazine hydrochloride, 2,4,6-trimethylphenyl hydrazine hydrochloride, hydrazine, 2,4,6-trimethylphenyl-, hydrochloride, mesityl hydrazine hydrochloride
(2,4,6-trimethylphenyl)hydrazine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only