Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trimethylphenyl isothiocyanate, 96%

Chemical Identifiers

Chemical Name or Material 2,4,6-Trimethylphenyl isothiocyanate
CAS 6095-82-5
Molecular Formula C10H11NS
Formula Weight 177.27
MDL Number MFCD00022055
UN Number UN2923
Beilstein 2690815
Synonym Mesityl isothiocyanate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1229403

  SDS
alfa aesar™
L1229403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1229406

  SDS
alfa aesar™
L1229406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 177.27
Molecular Formula C10H11NS
CAS 6095-82-5
UN Number UN2923
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Odor Irritating
Chemical Name or Material 2,4,6-Trimethylphenyl isothiocyanate
Assay 96%
MDL Number MFCD00022055
Beilstein 2690815
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 60° to 63°C
Quantity 1g
Synonym Mesityl isothiocyanate
Boiling Point 144° to 145°C (10mmHg)
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.