missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trimethylphenyl isothiocyanate, 96%

Manufacturer:  Alfa Aesar™ L1229403

 View more versions of this product

Catalog No. AAL1229403



Specifications

Specifications

2,4,6-Trimethylphenyl isothiocyanate
144°C to 145°C (10mmHg)
1g
C10H11NS
UN2923
2,4,6-trimethylphenyl isothiocyanate, mesityl isothiocyanate, 2,4,6-trimethylphenylisothiocyanate, 2-isothiocyanato-1,3,5-trimethyl-benzene, benzene, 2-isothiocyanato-1,3,5-trimethyl, 2,4,6-trimethylbenzenisothiocyanate, mesitylisothiocyanate, acmc-20aoen, isothiocyanic acid mesityl ester, #
CC1=CC(=C(C(=C1)C)N=C=S)C
177.265
177.27
Irritating
60°C to 63°C
6095-82-5
MFCD00022055
2690815
KKYMYPLVBCVDPL-UHFFFAOYSA-N
2-isothiocyanato-1,3,5-trimethylbenzene
138659
96%

Chemical Identifiers

6095-82-5
177.265
KKYMYPLVBCVDPL-UHFFFAOYSA-N
138659
CC1=CC(=C(C(=C1)C)N=C=S)C
C10H11NS
MFCD00022055
2,4,6-trimethylphenyl isothiocyanate, mesityl isothiocyanate, 2,4,6-trimethylphenylisothiocyanate, 2-isothiocyanato-1,3,5-trimethyl-benzene, benzene, 2-isothiocyanato-1,3,5-trimethyl, 2,4,6-trimethylbenzenisothiocyanate, mesitylisothiocyanate, acmc-20aoen, isothiocyanic acid mesityl ester, #
2-isothiocyanato-1,3,5-trimethylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

Documents