missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trimethylphenyl isocyanate, 98%

Manufacturer:  Alfa Aesar™ L1261103

 View more versions of this product

Catalog No. AAL1261103



Description

Description

Applications
2,4,6-Trimethylphenyl isocyanate is used as an organic chemical synthesis intermediate.

Solubility
It hydrolyzes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols, amines, bases and heat.
Specifications

Specifications

2,4,6-Trimethylphenyl isocyanate
Irritating
43°C to 46°C
2958-62-5
MFCD00019909
2085601
HJHZRZFONUPQAA-UHFFFAOYSA-N
2-isocyanato-1,3,5-trimethylbenzene
595239
98%
104°C (219°F)
218°C to 220°C
1g
C10H11NO
UN2811
2,4,6-trimethylphenyl isocyanate, mesityl isocyanate, 2,4,6-trimethylphenylisocyanate, acmc-1ckns, 2,4,6-trimethylbenzenisocyanate, 2-isocyanato-1,3,5-trimethyl-benzene, 2,4,6-trimethylphenyl isocyanate, #
CC1=CC(=C(C(=C1)C)N=C=O)C
161.204
161.21

Chemical Identifiers

2958-62-5
161.204
HJHZRZFONUPQAA-UHFFFAOYSA-N
595239
CC1=CC(=C(C(=C1)C)N=C=O)C
C10H11NO
MFCD00019909
2,4,6-trimethylphenyl isocyanate, mesityl isocyanate, 2,4,6-trimethylphenylisocyanate, acmc-1ckns, 2,4,6-trimethylbenzenisocyanate, 2-isocyanato-1,3,5-trimethyl-benzene, 2,4,6-trimethylphenyl isocyanate, #
2-isocyanato-1,3,5-trimethylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H334-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents