Learn More
Alfa Aesar™ 2,4,6-Trimethylphenol, 98%
Manufacturer: Alfa Aesar™ A1504122
Specifications
2,4,6-Trimethylphenol | |
70°C to 74°C | |
527-60-6 | |
MFCD00002235 | |
mesitol, phenol, 2,4,6-trimethyl, mesityl alcohol, 2-hydroxymesitylene, 2,4,6-trimetylofenol, 1,3,5-trimethylphenol, 2,4,6-trimethyl-phenol, 1-hydroxy-2,4,6-trimethylbenzene, 2,4,6-trimethylofenol, unii-fpz32614n6 | |
CC1=CC(=C(C(=C1)C)O)C | |
136.194 | |
136.2 |
220°C to 221°C | |
100g | |
C9H12O | |
UN2430 | |
BPRYUXCVCCNUFE-UHFFFAOYSA-N | |
2,4,6-trimethylphenol | |
10698 | |
98% |
Chemical Identifiers
527-60-6 | |
136.194 | |
BPRYUXCVCCNUFE-UHFFFAOYSA-N | |
10698 | |
CC1=CC(=C(C(=C1)C)O)C |
C9H12O | |
MFCD00002235 | |
mesitol, phenol, 2,4,6-trimethyl, mesityl alcohol, 2-hydroxymesitylene, 2,4,6-trimetylofenol, 1,3,5-trimethylphenol, 2,4,6-trimethyl-phenol, 1-hydroxy-2,4,6-trimethylbenzene, 2,4,6-trimethylofenol, unii-fpz32614n6 | |
2,4,6-trimethylphenol |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.
RTECSNumber : OX6590000
TSCA : TSCA