missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Trimethylphenethylalcohol, 99%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 35120-0050

 View more versions of this product

Catalog No. AC351200050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2, 4, 6-Trimethylphenethylalcohol
81° to 83°C
99%
98.5
Authentic
≥98.5%
C11H16O
164.25
06, IV, 3412
White
5g
6950-92-1
100.0
Mass or Fluffy Needles
99%
(CH3)3C6HCHCHOH
MFCD00020609

Chemical Identifiers

6950-92-1
100.0
C11H16O
164.25
2, 4, 6-Trimethylphenethylalcohol, 99%
98.5
99%
(CH3)3C6HCHCHOH
MFCD00020609
≥98.5%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 230-123-7

TSCA : TSCA