Please login to your online account to display your discounted pricing

2,4,6-Trimethylphenethylalcohol, 99%, Acros Organics


Quantity 1g

Manufacturer: thermo fisher scientific  35120-0010

 View more versions of this product

Catalog No. AC351200010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Mass or Fluffy Needles
Formula Weight 164.25
Quantity 1g
Color White
Beilstein 06, IV, 3412
TSCA TSCA
EINECS Number 230-123-7
Melting Point 81° to 83°C
CAS Max % 100.0
Infrared Spectrum Authentic
CAS Min % 98.5
CAS 6950-92-1
Chemical Name or Material 2, 4, 6-Trimethylphenethylalcohol
Name Note 99%
Molecular Formula C11H16O
MDL Number MFCD00020609
Assay Percent Range 99%
Linear Formula (CH3)3C6HCHCHOH

Specifications

Chemical Name or Material 2, 4, 6-Trimethylphenethylalcohol
Name Note 99%
CAS 6950-92-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11H16O
Linear Formula (CH3)3C6HCHCHOH
Formula Weight 164.25
MDL Number MFCD00020609