Please login to your online account to display your discounted pricing

2,4,6-Trimethylphenethylalcohol, 99%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2, 4, 6-Trimethylphenethylalcohol
Name Note 99%
CAS 6950-92-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11H16O
Linear Formula (CH3)3C6HCHCHOH
Formula Weight 164.25
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC351200050

  SDS
thermo fisher scientific
35120-0050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC351200010

  SDS
thermo fisher scientific
35120-0010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Mass or Fluffy Needles
Formula Weight 164.25
Quantity 5g
Color White
EINECS Number 230-123-7
TSCA TSCA
Melting Point 81° to 83°C
Beilstein 06, IV, 3412
CAS Max % 100.0
CAS Min % 98.5
Infrared Spectrum Authentic
Chemical Name or Material 2, 4, 6-Trimethylphenethylalcohol
Name Note 99%
Assay Percent Range 99%
Molecular Formula C11H16O
Linear Formula (CH3)3C6HCHCHOH
MDL Number MFCD00020609
CAS 6950-92-1