Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trimethylbenzyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 2,4,6-Trimethylbenzyl chloride
CAS 1585-16-6
Molecular Formula C10H13Cl
Formula Weight 168.67
MDL Number MFCD00000901
UN Number UN3261
Beilstein 386678
Synonym alpha(2)-Chloroisodurene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0042306

  SDS
alfa aesar™
L0042306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0042314

  SDS
alfa aesar™
L0042314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 168.67
Chemical Name or Material 2,4,6-Trimethylbenzyl chloride
Assay 98%
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
MDL Number MFCD00000901
CAS 1585-16-6
UN Number UN3261
Beilstein 386678
Synonym alpha(2)-Chloroisodurene
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Flash Point 106°C (222°F)
Odor Pungent
Boiling Point 114° to 115°C (10mmHg)
Melting Point 37° to 42°C
Quantity 5g
Molecular Formula C10H13Cl
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C