Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trimethylbenzyl alcohol, 98+%

$73.69 - $260.68

Chemical Identifiers

Chemical Name or Material 2,4,6-Trimethylbenzyl alcohol
CAS 4170-90-5
Molecular Formula C10H14O
Formula Weight 150.22
MDL Number MFCD00014422
Beilstein 1862608
Synonym 2-(Hydroxymethyl)mesitylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0195206

  SDS
alfa aesar™
L0195206
5g Each for $73.69

AAL0195214

  SDS
alfa aesar™
L0195214
25g Each for $260.68
Description & Specifications

Specifications

Formula Weight 150.22
Chemical Name or Material 2,4,6-Trimethylbenzyl alcohol
Molecular Formula C10H14O
Assay ≥98%
MDL Number MFCD00014422
Synonym 2-(Hydroxymethyl)mesitylene
Boiling Point 140°C (15mmHg)
Quantity 5g
Beilstein 1862608
RTECS Number DP1402000
Melting Point 85° to 89°C
CAS 4170-90-5