Learn More
2,4,6-Trimethylbenzoyl chloride, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC244280100
Specifications
2, 4, 6-Trimethylbenzoyl chloride | |
Glass bottle | |
143°C to 146°C (60.0mmHg) | |
10g | |
97.5 | |
97.5% min. (GC) | |
MFCD00013650 | |
UKRQMDIFLKHCRO-UHFFFAOYSA-N | |
2,4,6-trimethylbenzoyl chloride | |
97038 | |
Liquid |
Authentic | |
1.5280 to 1.53 | |
Yellow | |
938-18-1 | |
100.0 | |
C10H11ClO | |
2,4,6-trimethyl-benzoyl chloride, 2,4,6-trimethylbenzoyl cloride, 2,4,6-trimethylbenzoylchloride, 2,4,6trimethylbenzoyl chloride, 2-mesitylenecarbonyl chloride, acmc-209rno, benzoyl chloride, 2,4,6-trimethyl, ksc491c5b, mesitoyl chloride, pubchem13234 | |
CC1=CC(=C(C(=C1)C)C(=O)Cl)C | |
182.65 | |
182.65 | |
98% |
Chemical Identifiers
938-18-1 | |
182.65 | |
UKRQMDIFLKHCRO-UHFFFAOYSA-N | |
97038 | |
CC1=CC(=C(C(=C1)C)C(=O)Cl)C |
C10H11ClO | |
MFCD00013650 | |
2,4,6-trimethyl-benzoyl chloride, 2,4,6-trimethylbenzoyl cloride, 2,4,6-trimethylbenzoylchloride, 2,4,6trimethylbenzoyl chloride, 2-mesitylenecarbonyl chloride, acmc-209rno, benzoyl chloride, 2,4,6-trimethyl, ksc491c5b, mesitoyl chloride, pubchem13234 | |
2,4,6-trimethylbenzoyl chloride |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 213-339-6