missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Trimethylbenzoyl chloride, 98%, ACROS Organics™

$240.01 - $240.01

Chemical Identifiers

CAS 938-18-1
Molecular Formula C10H11ClO
Molecular Weight (g/mol) 182.65
MDL Number MFCD00013650
InChI Key UKRQMDIFLKHCRO-UHFFFAOYSA-N
Synonym 2,4,6-trimethyl-benzoyl chloride, 2,4,6-trimethylbenzoyl cloride, 2,4,6-trimethylbenzoylchloride, 2,4,6trimethylbenzoyl chloride, 2-mesitylenecarbonyl chloride, acmc-209rno, benzoyl chloride, 2,4,6-trimethyl, ksc491c5b, mesitoyl chloride, pubchem13234
PubChem CID 97038
IUPAC Name 2,4,6-trimethylbenzoyl chloride
SMILES CC1=CC(=C(C(=C1)C)C(=O)Cl)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC244280100
SDS View Documents
Thermo Fisher Scientific
AC244280100
10g Glass bottle
Each for $240.01
 
AC244280500
SDS View Documents
Thermo Fisher Scientific
AC244280500
50g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

938-18-1
182.65
UKRQMDIFLKHCRO-UHFFFAOYSA-N
97038
CC1=CC(=C(C(=C1)C)C(=O)Cl)C
C10H11ClO
MFCD00013650
2,4,6-trimethyl-benzoyl chloride, 2,4,6-trimethylbenzoyl cloride, 2,4,6-trimethylbenzoylchloride, 2,4,6trimethylbenzoyl chloride, 2-mesitylenecarbonyl chloride, acmc-209rno, benzoyl chloride, 2,4,6-trimethyl, ksc491c5b, mesitoyl chloride, pubchem13234
2,4,6-trimethylbenzoyl chloride

Specifications

Authentic
1.5280 to 1.53
Yellow
938-18-1
100.0
C10H11ClO
2,4,6-trimethyl-benzoyl chloride, 2,4,6-trimethylbenzoyl cloride, 2,4,6-trimethylbenzoylchloride, 2,4,6trimethylbenzoyl chloride, 2-mesitylenecarbonyl chloride, acmc-209rno, benzoyl chloride, 2,4,6-trimethyl, ksc491c5b, mesitoyl chloride, pubchem13234
CC1=CC(=C(C(=C1)C)C(=O)Cl)C
182.65
182.65
98%
Glass bottle
143°C to 146°C (60.0mmHg)
10g
97.5
97.5% min. (GC)
MFCD00013650
UKRQMDIFLKHCRO-UHFFFAOYSA-N
2,4,6-trimethylbenzoyl chloride
97038
Liquid
2, 4, 6-Trimethylbenzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 213-339-6

SDS