missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4,6-Trimethylbenzoic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1183130
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,4,6-Trimethylbenzoic acid | |
153°C to 155°C | |
MFCD00002481 | |
1866187 | |
FFFIRKXTFQCCKJ-UHFFFAOYSA-N | |
2,4,6-trimethylbenzoic acid | |
10194 | |
164.2 |
480-63-7 | |
C10H12O2 | |
250 g | |
mesitoic acid, mesitylenecarboxylic acid, beta-isodurylic acid, benzoic acid, 2,4,6-trimethyl, .beta.-isodurylic acid, unii-4pu2hg6787, 2-mesitylenecarboxylic acid, 2,4,6-trimethyl-benzoic acid, 2,6-trimethylbenzoic acid, benzoic acid,4,6-trimethyl | |
CC1=CC(C)=C(C(O)=O)C(C)=C1 | |
164.20 | |
CHEBI:64830 | |
99% |
Chemical Identifiers
480-63-7 | |
164.20 | |
FFFIRKXTFQCCKJ-UHFFFAOYSA-N | |
10194 | |
2,4,6-trimethylbenzoic acid |
C10H12O2 | |
MFCD00002481 | |
mesitoic acid, mesitylenecarboxylic acid, beta-isodurylic acid, benzoic acid, 2,4,6-trimethyl, .beta.-isodurylic acid, unii-4pu2hg6787, 2-mesitylenecarboxylic acid, 2,4,6-trimethyl-benzoic acid, 2,6-trimethylbenzoic acid, benzoic acid,4,6-trimethyl | |
CHEBI:64830 | |
CC1=CC(C)=C(C(O)=O)C(C)=C1 |
Safety and Handling
EINECSNumber : 207-553-9
RTECSNumber : DI0887010
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only