missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trimethylbenzoic acid, 99%

Manufacturer:  Alfa Aesar™ A1183118

 View more versions of this product

Catalog No. AAA1183118



Specifications

Specifications

2,4,6-Trimethylbenzoic acid
50g
C10H12O2
1866187
FFFIRKXTFQCCKJ-UHFFFAOYSA-N
2,4,6-trimethylbenzoic acid
10194
164.2
153°C to 155°C
480-63-7
MFCD00002481
mesitoic acid, mesitylenecarboxylic acid, beta-isodurylic acid, benzoic acid, 2,4,6-trimethyl, .beta.-isodurylic acid, unii-4pu2hg6787, 2-mesitylenecarboxylic acid, 2,4,6-trimethyl-benzoic acid, 2,6-trimethylbenzoic acid, benzoic acid,4,6-trimethyl
CC1=CC(=C(C(=C1)C)C(=O)O)C
164.204
CHEBI:64830
99%

Chemical Identifiers

480-63-7
164.204
FFFIRKXTFQCCKJ-UHFFFAOYSA-N
10194
2,4,6-trimethylbenzoic acid
C10H12O2
MFCD00002481
mesitoic acid, mesitylenecarboxylic acid, beta-isodurylic acid, benzoic acid, 2,4,6-trimethyl, .beta.-isodurylic acid, unii-4pu2hg6787, 2-mesitylenecarboxylic acid, 2,4,6-trimethyl-benzoic acid, 2,6-trimethylbenzoic acid, benzoic acid,4,6-trimethyl
CHEBI:64830
CC1=CC(=C(C(=C1)C)C(=O)O)C
Safety and Handling

Safety and Handling

RTECSNumber : DI0887010

TSCA : TSCA

SDS Documents