missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trimethylbenzeneboronic acid neopentyl glycol ester, 99%

$54.59 - $168.09

Chemical Identifiers

CAS 214360-78-8
Molecular Formula C14H21BO2
Molecular Weight (g/mol) 232.13
MDL Number MFCD01318178
InChI Key ITLDRNGUSKUMRG-UHFFFAOYSA-N
Synonym 2,4,6-trimethylbenzeneboronic acid neopentyl glycol ester, 5,5-dimethyl-2-2,4,6-trimethylphenyl-1,3,2-dioxaborinane, 2-mesityl-5,5-dimethyl-1,3,2-dioxaborinane, 2,4,6-trimethylbenzeneboronic acid neopentyl glycol cyclic ester, mesitylboronic acid 2,2-dimethyltrimethylene ester, 2,4,6-trimethyl-benzeneboronic acid neopentyl glycol cyclic ester, 2,4,6-trimethylphenylboronic acid neopentyl glycol ester, mesityleneboronic acid 2,2-dimethyltrimethylene cyclic ester
PubChem CID 11436188
IUPAC Name 5,5-dimethyl-2-(2,4,6-trimethylphenyl)-1,3,2-dioxaborinane
SMILES B1(OCC(CO1)(C)C)C2=C(C=C(C=C2C)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1723206 View Documents Alfa Aesar™
L1723206
5g Each for $168.09
 
AAL1723203 View Documents Alfa Aesar™
L1723203
1g Each for $54.59
 
Specifications

Chemical Identifiers

214360-78-8
232.13
ITLDRNGUSKUMRG-UHFFFAOYSA-N
11436188
B1(OCC(CO1)(C)C)C2=C(C=C(C=C2C)C)C
C14H21BO2
MFCD01318178
2,4,6-trimethylbenzeneboronic acid neopentyl glycol ester, 5,5-dimethyl-2-2,4,6-trimethylphenyl-1,3,2-dioxaborinane, 2-mesityl-5,5-dimethyl-1,3,2-dioxaborinane, 2,4,6-trimethylbenzeneboronic acid neopentyl glycol cyclic ester, mesitylboronic acid 2,2-dimethyltrimethylene ester, 2,4,6-trimethyl-benzeneboronic acid neopentyl glycol cyclic ester, 2,4,6-trimethylphenylboronic acid neopentyl glycol ester, mesityleneboronic acid 2,2-dimethyltrimethylene cyclic ester
5,5-dimethyl-2-(2,4,6-trimethylphenyl)-1,3,2-dioxaborinane

Specifications

46°C to 48°C
214360-78-8
MFCD01318178
2,4,6-trimethylbenzeneboronic acid neopentyl glycol ester, 5,5-dimethyl-2-2,4,6-trimethylphenyl-1,3,2-dioxaborinane, 2-mesityl-5,5-dimethyl-1,3,2-dioxaborinane, 2,4,6-trimethylbenzeneboronic acid neopentyl glycol cyclic ester, mesitylboronic acid 2,2-dimethyltrimethylene ester, 2,4,6-trimethyl-benzeneboronic acid neopentyl glycol cyclic ester, 2,4,6-trimethylphenylboronic acid neopentyl glycol ester, mesityleneboronic acid 2,2-dimethyltrimethylene cyclic ester
B1(OCC(CO1)(C)C)C2=C(C=C(C=C2C)C)C
232.13
232.13
2,4,6-Trimethylbenzeneboronic acid neopentyl glycol ester
5g
C14H21BO2
8689636
ITLDRNGUSKUMRG-UHFFFAOYSA-N
5,5-dimethyl-2-(2,4,6-trimethylphenyl)-1,3,2-dioxaborinane
11436188
99%