Please login to your online account to display your discounted pricing

2,4,6-Trimethylbenzene-1-thiol, 97%, Maybridge

$32.23 - $32.23

Chemical Identifiers

Chemical Name or Material 2, 4, 6-Trimethylbenzene-1-thiol
CAS 1541-10-2
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H12S
Linear Formula (CH3)3C6H2SH
Formula Weight 152.26
MDL Number MFCD00218475
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BTB12350EA

  SDS
fisher scientific company
BTB12350EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTB12350DA

  SDS
fisher scientific company
BTB12350DA
Amber Glass Bottle 1g Each for $32.23
Description & Specifications

Specifications

Formula Weight 152.26
Quantity 10g
Assay 97%
Density 1.0240g/mL
CAS Max % 100.0
Linear Formula (CH3)3C6H2SH
CAS Min % 96.5
MDL Number MFCD00218475
Packaging Amber Glass Bottle
Health Hazard 1 Warning
Assay Percent Range 97%
Molecular Formula C9H12S
CAS 1541-10-2
Chemical Name or Material 2, 4, 6-Trimethylbenzene-1-thiol
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.