Please login to your online account to display your discounted pricing

2,4,6-Trimethylbenzene-1-thiol, 97%, Maybridge


Quantity 25g

Manufacturer: fisher scientific company  BTB12350EB

 View more versions of this product

Catalog No. BTB12350EB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 152.26
Quantity 25g
Assay 97%
CAS Max % 100.0
Health Hazard 1 Warning
Linear Formula (CH3)3C6H2SH
Molecular Formula C9H12S
CAS Min % 96.5
MDL Number MFCD00218475
CAS 1541-10-2
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Assay Percent Range 97%
Chemical Name or Material 2, 4, 6-Trimethylbenzene-1-thiol
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2, 4, 6-Trimethylbenzene-1-thiol
CAS 1541-10-2
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H12S
Linear Formula (CH3)3C6H2SH
Formula Weight 152.26
MDL Number MFCD00218475
Accessories

Warning

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

SDS & Safety Documents