missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',4',6'-Trimethylacetophenone, 98%

Manufacturer:  Alfa Aesar™ A1154922

 View more versions of this product

Catalog No. AAA1154922


 

Description

Description

Applications
2′,4′,6′-Trimethylacetophenone is used as laboratory chemicals, manufacture of substances and intermediated for chemical synthesis.

Solubility
Difficult to mix.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Specifications

2',4',6'-Trimethylacetophenone
235°C to 236°C
1667-01-2
C11H14O
2044595
XWCIICLTKWRWCI-UHFFFAOYSA-N
1-(2,4,6-trimethylphenyl)ethanone
15461
>110°C (230°F)
100g
98%
MFCD00008735
2-Acetylmesitylene
CC1=CC(=C(C(=C1)C)C(=O)C)C
162.232
162.23

Chemical Identifiers

1667-01-2
162.232
XWCIICLTKWRWCI-UHFFFAOYSA-N
15461
C11H14O
MFCD00008735
2-Acetylmesitylene
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents