Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,6-Trimethyl-N-methylaniline, 97%

Chemical Identifiers

Chemical Name or Material 2,4,6-Trimethyl-N-methylaniline
CAS 13021-14-2
Formula Weight 149.24
MDL Number MFCD06200698
UN Number UN3082
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5515103

  SDS
alfa aesar™
H5515103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5515106

  SDS
alfa aesar™
H5515106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55151MD

  SDS
alfa aesar™
H55151MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 149.24
Chemical Name or Material 2,4,6-Trimethyl-N-methylaniline
Refractive Index 1.5320
Physical Form Liquid
Assay 97%
MDL Number MFCD06200698
CAS 13021-14-2
UN Number UN3082
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
Density 0.935g/mL
Boiling Point 229°C
Color Colorless to brown
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.