missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Trimethyl-N-[4-(trifluoromethyl)benzyl]benzenesulfonamide, 97%, Alfa Aesar™

$171.97 - $602.70

Chemical Identifiers

CAS 305849-16-5
Molecular Formula C17H18F3NO2S
Molecular Weight (g/mol) 357.391
MDL Number MFCD01652040
InChI Key FVZQXGUIEZGZJJ-UHFFFAOYSA-N
Synonym 2,4,6-trimethyl-n-4-trifluoromethyl benzyl benzenesulfonamide
PubChem CID 3700146
IUPAC Name 2,4,6-trimethyl-N-[[4-(trifluoromethyl)phenyl]methyl]benzenesulfonamide
SMILES CC1=CC(=C(C(=C1)C)S(=O)(=O)NCC2=CC=C(C=C2)C(F)(F)F)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH55384MD View Documents Thermo Fisher Scientific
H55384MD
250mg Each for $171.97
 
AAH5538403 View Documents Thermo Fisher Scientific
H5538403
1g Each for $602.70
 
Description

Description

Notes
Reacts with strong oxidizing agents.
Specifications

Chemical Identifiers

305849-16-5
357.391
FVZQXGUIEZGZJJ-UHFFFAOYSA-N
3700146
CC1=CC(=C(C(=C1)C)S(=O)(=O)NCC2=CC=C(C=C2)C(F)(F)F)C
C17H18F3NO2S
MFCD01652040
2,4,6-trimethyl-n-4-trifluoromethyl benzyl benzenesulfonamide
2,4,6-trimethyl-N-[[4-(trifluoromethyl)phenyl]methyl]benzenesulfonamide

Specifications

250mg
C17H18F3NO2S
21328032
FVZQXGUIEZGZJJ-UHFFFAOYSA-N
2,4,6-trimethyl-N-[[4-(trifluoromethyl)phenyl]methyl]benzenesulfonamide
3700146
97%
305849-16-5
MFCD01652040
2,4,6-trimethyl-n-4-trifluoromethyl benzyl benzenesulfonamide
CC1=CC(=C(C(=C1)C)S(=O)(=O)NCC2=CC=C(C=C2)C(F)(F)F)C
357.391
357.39
2,4,6-Trimethyl-N-[4-(trifluoromethyl)benzyl]benzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures