missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Trimethyl-N-[3-(trifluoromethyl)phenyl]benzenesulfonamide, Alfa Aesar™

Activates phospholipase C

$128.62 - $521.24

Chemical Identifiers

CAS 200933-14-8
Molecular Formula C16H16F3NO2S
Molecular Weight (g/mol) 343.364
MDL Number MFCD00095824
InChI Key ZIIUUSVHCHPIQD-UHFFFAOYSA-N
Synonym m-3m3fbs, 2,4,6-trimethyl-n-3-trifluoromethyl phenyl benzenesulfonamide, m3m3fbs, 2,4,6-trimethyl-n-3-trifluoromethyl phenyl benzene-1-sulfonamide, 2,4,6-trimethyl-n-3-trifluoromethyl-phenyl-benzenesulfonamide, phospholipase c activator, m-3m3fbs, n-3-trifluoromethylphenyl-2,4,6-trimethylbenzenesulfonamide, 2,4,6-trimethyl-n-3-trifluoromethyl phenyl benzenesulfonamide, 2,4,6-trimethyl-n-m-3-trifluoromethylphenyl benzenesulfonamide, tocris-1941
PubChem CID 761523
IUPAC Name 2,4,6-trimethyl-N-[3-(trifluoromethyl)phenyl]benzenesulfonamide
SMILES CC1=CC(=C(C(=C1)C)S(=O)(=O)NC2=CC=CC(=C2)C(F)(F)F)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ65999MA View Documents Alfa Aesar
J65999MA
10mg Each for $128.62
 
AAJ65999MF View Documents Alfa Aesar
J65999MF
50mg Each for $521.24
 
Specifications

Chemical Identifiers

200933-14-8
343.364
ZIIUUSVHCHPIQD-UHFFFAOYSA-N
761523
CC1=CC(=C(C(=C1)C)S(=O)(=O)NC2=CC=CC(=C2)C(F)(F)F)C
C16H16F3NO2S
MFCD00095824
m-3m3fbs, 2,4,6-trimethyl-n-3-trifluoromethyl phenyl benzenesulfonamide, m3m3fbs, 2,4,6-trimethyl-n-3-trifluoromethyl phenyl benzene-1-sulfonamide, 2,4,6-trimethyl-n-3-trifluoromethyl-phenyl-benzenesulfonamide, phospholipase c activator, m-3m3fbs, n-3-trifluoromethylphenyl-2,4,6-trimethylbenzenesulfonamide, 2,4,6-trimethyl-n-3-trifluoromethyl phenyl benzenesulfonamide, 2,4,6-trimethyl-n-m-3-trifluoromethylphenyl benzenesulfonamide, tocris-1941
2,4,6-trimethyl-N-[3-(trifluoromethyl)phenyl]benzenesulfonamide

Specifications

White
10mg
C16H16F3NO2S
m-3m3fbs, 2,4,6-trimethyl-n-3-trifluoromethyl phenyl benzenesulfonamide, m3m3fbs, 2,4,6-trimethyl-n-3-trifluoromethyl phenyl benzene-1-sulfonamide, 2,4,6-trimethyl-n-3-trifluoromethyl-phenyl-benzenesulfonamide, phospholipase c activator, m-3m3fbs, n-3-trifluoromethylphenyl-2,4,6-trimethylbenzenesulfonamide, 2,4,6-trimethyl-n-3-trifluoromethyl phenyl benzenesulfonamide, 2,4,6-trimethyl-n-m-3-trifluoromethylphenyl benzenesulfonamide, tocris-1941
CC1=CC(=C(C(=C1)C)S(=O)(=O)NC2=CC=CC(=C2)C(F)(F)F)C
343.364
343.36
2,4,6-Trimethyl-N-[3-(trifluoromethyl)phenyl]benzenesulfonamide
114°C to 118°C
200933-14-8
MFCD00095824
ZIIUUSVHCHPIQD-UHFFFAOYSA-N
2,4,6-trimethyl-N-[3-(trifluoromethyl)phenyl]benzenesulfonamide
761523
Powder
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning