Please login to your online account to display your discounted pricing

2,4,6-Trimethoxypyrimidine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4, 6-Trimethoxypyrimidine
Assay Percent Range 98%
Molecular Formula C7H10N2O3
Formula Weight 170.17
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC426740010

 
thermo fisher scientific
AC426740010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC426742500

 
thermo fisher scientific
AC426742500
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 170.17
Quantity 1g
Chemical Name or Material 2, 4, 6-Trimethoxypyrimidine
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C7H10N2O3
CAS 13106-85-9