missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Trimethoxyboroxin, TCI America™

Manufacturer:  TCI America T158125G

 View more versions of this product

Catalog No. T158125G



Specifications

Specifications

2,4,6-Trimethoxyboroxin
25g
C3H9B3O6
2924
ZFMOJHVRFMOIGF-UHFFFAOYSA-N
2,4,6-trimethoxy-1,3,5,2,4,6-trioxatriborinane
66880
Clear Liquid at 20°C
Yellow
102-24-9
MFCD00005946
trimethoxyboroxine, trimethoxyboroxin, boroxin, trimethoxy, trimethoxycyclotriboroxane, 2,4,6-trimethoxyboroxin, triboron trimethyl hexaoxide, boroxin, 2,4,6-trimethoxy, boric acid h3b3o6 , trimethyl ester, trimethoxy-boroxi, trimethoxyboroxole
B1(OB(OB(O1)OC)OC)OC
173.529
173.53
≥96.0% (T)

Chemical Identifiers

102-24-9
173.529
ZFMOJHVRFMOIGF-UHFFFAOYSA-N
66880
B1(OB(OB(O1)OC)OC)OC
C3H9B3O6
MFCD00005946
trimethoxyboroxine, trimethoxyboroxin, boroxin, trimethoxy, trimethoxycyclotriboroxane, 2,4,6-trimethoxyboroxin, triboron trimethyl hexaoxide, boroxin, 2,4,6-trimethoxy, boric acid h3b3o6 , trimethyl ester, trimethoxy-boroxi, trimethoxyboroxole
2,4,6-trimethoxy-1,3,5,2,4,6-trioxatriborinane
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3868

RTECSNumber : ED8720000

TSCA : Yes

Documents