missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4,6-Trimethoxybenzylamine hydrochloride, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 146548-59-6 |
---|---|
Molecular Formula | C10H15NO3·ClH |
Molecular Weight (g/mol) | 233.7 |
InChI Key | BLFRMOOGAICNSZ-UHFFFAOYSA-N |
Synonym | 2,4,6-trimethoxybenzylamine hydrochloride, 2,4,6-trimethoxyphenyl methanamine hydrochloride, 1-2,4,6-trimethoxyphenyl methanamine hydrochloride, pubchem7389, 2, 4, 6-trimethoxybenzylamine hydrochloride, acmc-1c5nr, benzenemethanamine, 2,4,6-trimethoxy-, hydrochloride, 1-2,4,6-trimethoxyphenyl methanamine-hydrogen chloride 1/1 |
PubChem CID | 16211914 |
IUPAC Name | (2,4,6-trimethoxyphenyl)methanamine;hydrochloride |
SMILES | COC1=CC(=C(C(=C1)OC)CN)OC.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC437330010
|
Thermo Scientific Chemicals
437330010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
146548-59-6 | |
233.7 | |
2,4,6-trimethoxybenzylamine hydrochloride, 2,4,6-trimethoxyphenyl methanamine hydrochloride, 1-2,4,6-trimethoxyphenyl methanamine hydrochloride, pubchem7389, 2, 4, 6-trimethoxybenzylamine hydrochloride, acmc-1c5nr, benzenemethanamine, 2,4,6-trimethoxy-, hydrochloride, 1-2,4,6-trimethoxyphenyl methanamine-hydrogen chloride 1/1 | |
(2,4,6-trimethoxyphenyl)methanamine;hydrochloride |
C10H15NO3·ClH | |
BLFRMOOGAICNSZ-UHFFFAOYSA-N | |
16211914 | |
COC1=CC(=C(C(=C1)OC)CN)OC.Cl |
Specifications
146548-59-6 | |
100.0 | |
97.5% min. (HPLC) | |
C10H15NO3·ClH | |
2,4,6-trimethoxybenzylamine hydrochloride, 2,4,6-trimethoxyphenyl methanamine hydrochloride, 1-2,4,6-trimethoxyphenyl methanamine hydrochloride, pubchem7389, 2, 4, 6-trimethoxybenzylamine hydrochloride, acmc-1c5nr, benzenemethanamine, 2,4,6-trimethoxy-, hydrochloride, 1-2,4,6-trimethoxyphenyl methanamine-hydrogen chloride 1/1 | |
COC1=CC(=C(C(=C1)OC)CN)OC.Cl | |
233.7 | |
233.7 | |
2, 4, 6-Trimethoxybenzylamine hydrochloride |
97.5 | |
Authentic | |
Glass bottle | |
1g | |
BLFRMOOGAICNSZ-UHFFFAOYSA-N | |
(2,4,6-trimethoxyphenyl)methanamine;hydrochloride | |
16211914 | |
98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
GHS Signal Word: Warning
RUO â Research Use Only