missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Trimethoxybenzylamine hydrochloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC437330050

 View more versions of this product

Catalog No. AC437330050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 4, 6-Trimethoxybenzylamine hydrochloride
Glass bottle
204°C to 207°C
146548-59-6
100.0
C10H15NO3·ClH
BLFRMOOGAICNSZ-UHFFFAOYSA-N
(2,4,6-trimethoxyphenyl)methanamine;hydrochloride
16211914
Powder
Authentic
White
5g
97.5
97.5% min. (HPLC)
1-2,4,6-trimethoxyphenyl methanamine hydrochloride, 1-2,4,6-trimethoxyphenyl methanamine-hydrogen chloride 1/1, 2, 4, 6-trimethoxybenzylamine hydrochloride, 2,4,6-trimethoxybenzylamine hydrochloride, 2,4,6-trimethoxyphenyl methanamine hydrochloride, acmc-1c5nr, benzenemethanamine, 2,4,6-trimethoxy-, hydrochloride, pubchem7389
COC1=CC(=C(C(=C1)OC)CN)OC.Cl
233.7
233.7
98%

Chemical Identifiers

146548-59-6
233.7
1-2,4,6-trimethoxyphenyl methanamine hydrochloride, 1-2,4,6-trimethoxyphenyl methanamine-hydrogen chloride 1/1, 2, 4, 6-trimethoxybenzylamine hydrochloride, 2,4,6-trimethoxybenzylamine hydrochloride, 2,4,6-trimethoxyphenyl methanamine hydrochloride, acmc-1c5nr, benzenemethanamine, 2,4,6-trimethoxy-, hydrochloride, pubchem7389
(2,4,6-trimethoxyphenyl)methanamine;hydrochloride
C10H15NO3·ClH
BLFRMOOGAICNSZ-UHFFFAOYSA-N
16211914
COC1=CC(=C(C(=C1)OC)CN)OC.Cl
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.

GHS Signal Word: Warning

SDS Documents