missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4,6-Trimethoxybenzylamine hydrochloride, 98%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC437330010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 4, 6-Trimethoxybenzylamine hydrochloride | |
97.5 | |
White | |
97.5% min. (HPLC) | |
C10H15NO3·ClH | |
1-2,4,6-trimethoxyphenyl methanamine hydrochloride, 1-2,4,6-trimethoxyphenyl methanamine-hydrogen chloride 1/1, 2, 4, 6-trimethoxybenzylamine hydrochloride, 2,4,6-trimethoxybenzylamine hydrochloride, 2,4,6-trimethoxyphenyl methanamine hydrochloride, acmc-1c5nr, benzenemethanamine, 2,4,6-trimethoxy-, hydrochloride, pubchem7389 | |
BLFRMOOGAICNSZ-UHFFFAOYSA-N | |
(2,4,6-trimethoxyphenyl)methanamine;hydrochloride | |
16211914 | |
Powder |
146548-59-6 | |
100.0 | |
204°C to 207°C | |
Authentic | |
Glass bottle | |
1g | |
COC1=CC(=C(C(=C1)OC)CN)OC.Cl | |
233.7 | |
233.7 | |
98% |
Chemical Identifiers
146548-59-6 | |
233.7 | |
1-2,4,6-trimethoxyphenyl methanamine hydrochloride, 1-2,4,6-trimethoxyphenyl methanamine-hydrogen chloride 1/1, 2, 4, 6-trimethoxybenzylamine hydrochloride, 2,4,6-trimethoxybenzylamine hydrochloride, 2,4,6-trimethoxyphenyl methanamine hydrochloride, acmc-1c5nr, benzenemethanamine, 2,4,6-trimethoxy-, hydrochloride, pubchem7389 | |
(2,4,6-trimethoxyphenyl)methanamine;hydrochloride |
C10H15NO3·ClH | |
BLFRMOOGAICNSZ-UHFFFAOYSA-N | |
16211914 | |
COC1=CC(=C(C(=C1)OC)CN)OC.Cl |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
GHS Signal Word: Warning
RUO – Research Use Only