Learn More
2,4,6-Trimethoxybenzonitrile, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2354014
Description
2,4,6-Trimethoxybenzonitrile is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4,6-Trimethoxybenzonitrile is used as pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications
2,4,6-Trimethoxybenzonitrile | |
140°C to 143°C | |
C10H11NO3 | |
25 g | |
Insoluble in water. | |
COC1=CC(=C(C(=C1)OC)C#N)OC | |
193.202 | |
193.2 |
2571-54-2 | |
Characteristic | |
MFCD00001787 | |
benzonitrile, 2,4,6-trimethoxy, 2,4,6-trimethoxy-benzonitrile, 2,4,6-trimethoxybenzenecarbonitrile, acmc-20aoeu, 2,4,6-trimethoxy benzonitrile, benzonitrile,2,4,6-trimethoxy, 2,4,6-trimethoxybenzonitrile | |
GBRHJUMDNWLSCT-UHFFFAOYSA-N | |
2,4,6-trimethoxybenzonitrile | |
75731 | |
98% |
Chemical Identifiers
2571-54-2 | |
193.202 | |
GBRHJUMDNWLSCT-UHFFFAOYSA-N | |
2,4,6-trimethoxybenzonitrile |
C10H11NO3 | |
MFCD00001787 | |
75731 | |
COC1=CC(=C(C(=C1)OC)C#N)OC |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
H301+H311+H331
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 219-917-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only