missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Trimethoxybenzaldehyde 98%, ACROS Organics™

Manufacturer:  Acros Organics 160530010

 View more versions of this product

Catalog No. AC160530010



Specifications

Specifications

2, 4, 6-Trimethoxybenzaldehyde
Glass bottle
118.0°C to 120.0°C
98%
97.5
98%
(CH3O)3C6H2CHO
benzaldehyde, 2,4,6-trimethoxy, 2,4,6-trimethoxybenz aldehyde, pubchem8264, acmc-1cuhn, ksc490m2h, 2,4,6-trimethoxy-benzaldehyde, 2,4,6-trimethoxybenz-aldehyde, 2,4,6-trimethoxy benz aldehyde, crbzvdlxaiferf-uhfffaoysa, 1-formyl-2,4,6-trimethoxybenzene
COC1=CC(=C(C(=C1)OC)C=O)OC
196.202
196.2
98%
Authentic
Beige
1g
830-79-5
100.0
C10H12O4
MFCD00003313
CRBZVDLXAIFERF-UHFFFAOYSA-N
2,4,6-trimethoxybenzaldehyde
70019
Crystalline Powder

Chemical Identifiers

830-79-5
196.202
CRBZVDLXAIFERF-UHFFFAOYSA-N
70019
COC1=CC(=C(C(=C1)OC)C=O)OC
C10H12O4
MFCD00003313
benzaldehyde, 2,4,6-trimethoxy, 2,4,6-trimethoxybenz aldehyde, pubchem8264, acmc-1cuhn, ksc490m2h, 2,4,6-trimethoxy-benzaldehyde, 2,4,6-trimethoxybenz-aldehyde, 2,4,6-trimethoxy benz aldehyde, crbzvdlxaiferf-uhfffaoysa, 1-formyl-2,4,6-trimethoxybenzene
2,4,6-trimethoxybenzaldehyde
Safety and Handling

Safety and Handling

EINECSNumber : 212-598-2

RTECSNumber : CU8460200

TSCA : TSCA

SDS Documents