Please login to your online account to display your discounted pricing

2,4,6-Trimethoxybenzaldehyde 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4, 6-Trimethoxybenzaldehyde
Name Note 98%
CAS 830-79-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H12O4
Linear Formula (CH3O)3C6H2CHO
Formula Weight 196.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC160530010

  SDS
thermo fisher scientific
160530010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC160530250

  SDS
thermo fisher scientific
160530250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC160532500

  SDS
thermo fisher scientific
AC160532500
Glass bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 196.2
Quantity 1g
Color Beige
TSCA TSCA
EINECS Number 212-598-2
Melting Point 118° to 120°C
RTECS Number CU8460200
CAS Max % 100.0
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 830-79-5
Molecular Formula C10H12O4
MDL Number MFCD00003313
Assay Percent Range 98%
Name Note 98%
Chemical Name or Material 2, 4, 6-Trimethoxybenzaldehyde
Linear Formula (CH3O)3C6H2CHO